Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Two Shelf Utility Cart

Rubbermaid® commercial heavy-duty utility cart, PP, 500 lbs., 115.0 cm (45.2"), 65.7 cm (25.88"), 84.3 cm (33.2")
  470006-162
 :  RCP452088BK
Two Shelf Utility Cart
  470006-162
Two Shelf Utility Cart
Two Shelf Utility Cart

Rubbermaid® commercial heavy-duty utility cart, PP, 500 lbs., 115.0 cm (45.2"), 65.7 cm (25.88"), 84.3 cm (33.2")

 
$11,330.68
   
Each (1UN)
 

Perfect for transporting materials, supplies, and heavy loads in almost any environment.

  • Ergonomically designed push handle
  • Molded-in storage compartments
  • Easy-access bottomshelf

Handy, durable utility cart features rust-, dent,- chip- and peel-proof structural foam construction. Ergonomically designed push handle provides optimum hand placement and push height improving control and worker safety. Molded-in storage compartments, holsters, hooks and notches. Rounded corners and shelf walls won't damage furniture and walls. Variable grip-height handle improves control and worker safety. Built-in holster and molded-in fittings allow for easy addition of pipes or rods. Standard color is black.

 
  • Width:
    65.7 cm (25.88")
  • Description:
    Rubbermaid® commercial heavy-duty utility cart
  • Height:
    84.3 cm (33.2")
  • Length:
    115.0 cm (45.2")
  • Material:
    PP
  • Load Capacity:
    500 lbs.