Ohaus® Spring Scales
8002-MA - 20 g (0.50 oz.) graduation intervals, 500 g (18 oz.)
470006-148
: 80000041
These spring scales are designed to stand up to heavy student use.
- Scales withstand sudden load release
- Individually calibrated
- Color-coded
- High-contrast markings
Each scale has been individually calibrated for accuracy and can be fully zeroed. They withstand sudden load releases without breaking.
Scales are color coded, and high-contrast markings make readings easy.
- Description:8002-MA - 20 g (0.50 oz.) graduation intervals
- Weighing Capacity:500 g (18 oz.)