Orders LinkContactUsLinkComponent
Ohaus® Spring Scales
8002-MA - 20 g (0.50 oz.) graduation intervals, 500 g (18 oz.)
  470006-148
 :  80000041
Ohaus® Spring Scales
  470006-148
Ohaus® Spring Scales
Ohaus® Spring Scales
8002-MA - 20 g (0.50 oz.) graduation intervals, 500 g (18 oz.)
$1,026.04
Each (1UN)
These spring scales are designed to stand up to heavy student use.
  • Scales withstand sudden load release
  • Individually calibrated
  • Color-coded
  • High-contrast markings
Each scale has been individually calibrated for accuracy and can be fully zeroed. They withstand sudden load releases without breaking.
Scales are color coded, and high-contrast markings make readings easy.
  • Description:
    8002-MA - 20 g (0.50 oz.) graduation intervals
  • Weighing Capacity:
    500 g (18 oz.)