Specific Gravity Hydrometer
Specific Gravity Hydrometer, 0.70–1.00, 0.005 divisions
Catalog # 470005-562
Supplier: G&W INSTRUMENTS
Número de proveedor #: A-1177
Jump to:
Product Details & Documents
Use these hydrometers with all processes where specific gravity or density of liquids is an important control.
The design of this hydrometer ensures easy cleaning and a low likelihood of breakage. The clear markings allow accurate readings. Accuracy: ±0.001. Size: 12"L.
Especificaciones
- Description:Specific Gravity Hydrometer, 0.70–1.00, 0.005 divisions