Orders LinkContactUsLinkComponent
Cylindrical Battery Jars
Cylindrical Battery Jars
Cylindrical Battery Jars, 4 L
Catalog # 470005-290
Supplier:   VARIETY GLASS INC
Número de proveedor #:  618-3
Cylindrical Battery Jars
Catalog # 470005-290
Supplier:  VARIETY GLASS INC
Cylindrical Battery Jars
Cylindrical Battery Jars
Cylindrical Battery Jars, 4 L
Price
$12,408.43
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Ideal for a Variety of Uses.
  • Heavy walls
  • Ground and beveled top edges
Use for aquarium, terrarium, water baths, or for specific gravity experiments.
Especificaciones
  • Capacity:
    4 L
  • Description:
    Cylindrical Battery Jars