Ohaus® Spring Scales
8002-PN - 0.04 lbs. (0.2 N) graduation intervals, 1.12 lbs. (5 N)
470004-632
: 80000042
These spring scales are designed to stand up to heavy student use.
- Scales withstand sudden load release
- Individually calibrated
- Color-coded
- High-contrast markings
Each scale has been individually calibrated for accuracy and can be fully zeroed. They withstand sudden load releases without breaking.
Scales are color coded, and high-contrast markings make readings easy.
- Description:8002-PN - 0.04 lbs. (0.2 N) graduation intervals
- Weighing Capacity:1.12 lbs. (5 N)