Orders LinkContactUsLinkComponent
Ohaus® Spring Scales
8002-PN - 0.04 lbs. (0.2 N) graduation intervals, 1.12 lbs. (5 N)
  470004-632
 :  80000042
Ohaus® Spring Scales
  470004-632
Ohaus® Spring Scales
Ohaus® Spring Scales
8002-PN - 0.04 lbs. (0.2 N) graduation intervals, 1.12 lbs. (5 N)
$1,114.00
Each (1UN)
These spring scales are designed to stand up to heavy student use.
  • Scales withstand sudden load release
  • Individually calibrated
  • Color-coded
  • High-contrast markings
Each scale has been individually calibrated for accuracy and can be fully zeroed. They withstand sudden load releases without breaking.
Scales are color coded, and high-contrast markings make readings easy.
  • Description:
    8002-PN - 0.04 lbs. (0.2 N) graduation intervals
  • Weighing Capacity:
    1.12 lbs. (5 N)