United Scientific™ Pull-Type Spring Scales (Metric/Newton)
Spring scale, green, 500 g/5 N, graduation interval: 0.1 N
470000-124
: SB0500-P
With these scales, students see the spring action as it occurs.
- Color-coded
- Clear plastic tube
- Zero adjustment
These color-coded spring scales have a clear-tube design that allows students to see the spring action as it occurs. Each scale is 7 inches long, and features zero adjustment. The set of six comes with one of each weight capacity.
: Caution:WARNING: CHOKING HAZARD This product contains small parts is not for children under three years.
- Description:Spring scale, green
- Weighing Capacity:500 g/5 N, graduation interval: 0.1 N