Orders LinkContactUsLinkComponent
pH-Indicator Paper, MilliporeSigma
pH-Indicator Paper, MilliporeSigma
pH-Indicator Paper, Congo Red Paper Roll with Color Scale, 3 rolls, 4.8 m, —, <3 blue-violet/>5 red-orange
Catalog # EM1.09514.0003
Supplier #:  1095140003
pH-Indicator Paper, MilliporeSigma
Catalog # EM1.09514.0003
pH-Indicator Paper, MilliporeSigma
pH-Indicator Paper, Congo Red Paper Roll with Color Scale, 3 rolls, 4.8 m, —, <3 blue-violet/>5 red-orange
Price
$2,528.28
Log in for availability
Each (1UN) Case de 100 (1UN)
Product Details & Documents
pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences
MilliporeSigma # 1.09562.0003 replaces and is identical to MilliporeSigma # 9562-2.
MilliporeSigma # 1.09564.0003 replaces and is identical to MilliporeSigma # 9564-2.
pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. Therefore, the paper is dryed, cut and packed into the corresponding sizes.
Specifications
  • Description:
    pH-Indicator Paper, Congo Red Paper Roll with Color Scale
  • Length:
    4.8 m
  • Size:
    3 rolls
  • pH Range:
    <3 blue-violet/>5 red-orange
  • pH Graduation: