pH-Indicator Paper, MilliporeSigma
pH-Indicator Paper, Congo Red Paper Roll with Color Scale, 3 rolls, 4.8 m, —, <3 blue-violet/>5 red-orange
Supplier: MERCK, S.A. DE C.V. SV
Supplier #: 1095140003
Jump to:
Product Details & Documents
pH indicator papers supplied in rolls are better suited for longterm storage, since they are protected against external influences
MilliporeSigma # 1.09562.0003 replaces and is identical to MilliporeSigma # 9562-2.
MilliporeSigma # 1.09564.0003 replaces and is identical to MilliporeSigma # 9564-2.
pH indicator papers are manufactured by impregnating high-quality filter paper with indicator or mixed-indicator solutions. Therefore, the paper is dryed, cut and packed into the corresponding sizes.
Documents
Specifications
- Description:pH-Indicator Paper, Congo Red Paper Roll with Color Scale
- Length:4.8 m
- Size:3 rolls
- pH Range:<3 blue-violet/>5 red-orange
- pH Graduation:—