PLC Plates, MilliporeSigma
Product Details & Documents
PLC plates allow users to separate samples that vary greatly in size - from grams down to milligrams
Product # 1.05434.0001 replaces and is identical to Product # 5434-7.
Product # 1.05637.0001 replaces and is identical to Product # 5637-7.
Product # 1.05717.0001 replaces and is identical to Product # 5717-7.
Product # 1.05726.0001 replaces and is identical to Product # 5726-7.
Product # 1.05744.0001 replaces and is identical to Product # 5744-7.
Product # 1.05745.0001 replaces and is identical to Product # 5745-7.
Product # 1.13894.0001 replaces and is identical to Product # 13894-7.
Product # 1.13895.0001 replaces and is identical to Product # 13895-7.
Available with or without a fluorescence indicator. They also use the same proven silica-binder technology as in analytical TLC plates. To isolate the substance by extraction, users can simply scrape the spot from the layer.
Especificaciones
- Coating material:Silica gel
- Layer thickness:2000 µm
- Plate size:200×200 mm
- Pore size:60 Å
- Fluorescent Indicator:No
- No. of Plates:12
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



