Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

3" Test Sieves, W.S. Tyler™

3", 44.5 mm (1¾"), Stainless, 1 mm, #18, Stainless steel
  30624-142
 :   W.S. Tyler
 :  1295
3" Test Sieves, W.S. Tyler™
  30624-142
3" Test Sieves, W.S. Tyler™
3" Test Sieves, W.S. Tyler™

3", 44.5 mm (1¾"), Stainless, 1 mm, #18, Stainless steel

 
$2,983.50
   
Each (1UN)
 

These test sieves are available with stainless or brass frame and stainless cloth.

  • Depth to cloth: 1-1/4"
 
  • Ø:
    3"
  • Height:
    44.5 mm (1¾")
  • Mesh Material:
    Stainless
  • Mesh Size:
    1 mm
  • USA Series:
    #18
  • Frame Material:
    Stainless steel