Orders LinkContactUsLinkComponent
Switch-Grid™ Test Tube Rack, Rack Only
Switch-Grid™ Test Tube Rack, Rack Only
Set of Grids, Fuchsia, 20 - 25 mm
Catalog # 89260-610
Switch-Grid™ Test Tube Rack, Rack Only
Catalog # 89260-610
Supplier:  
Switch-Grid™ Test Tube Rack, Rack Only
Switch-Grid™ Test Tube Rack, Rack Only
Set of Grids, Fuchsia, 20 - 25 mm
Discontinued
Product Details & Documents
The innovative Switch-Grid™ rack utilizes interchangeable grids for ultimate flexibility. This split-frame rack holds up to two different tube sizes at the same time and allows users to quickly switch grids to hold other sizes as needed.
  • 10 to 30 mm diameter test tubes fit in 5 colored-coded grid inserts
  • Can be easily cleaned with common lab detergents, ethanol, or isopropyl alcohol
  • Three-tier design securely holds tubes in place while non-slip rubber feet prevent sliding on the lab bench
  • Rack available without grids for use with spare grids
This rack is the smart, adaptable solution for optimizing test tube organization and streamlining laboratory workflows. Constructed from chemical-resistant polyoxymethylene plastic, both the frame and grids are autoclavable at 121 °C (250 °F).
Especificaciones
  • Description:
    Set of Grids, Fuchsia
  • For Tube O.D.:
    20 - 25 mm