Orders LinkContactUsLinkComponent
SmartCal™ Moisture Analyzer Test Substance
SmartCal™ Moisture Analyzer Test Substance
Moisture analyzer test system (set of 12)
Catalog # 89237-464
Número de proveedor #:  30005792
SmartCal™ Moisture Analyzer Test Substance
Catalog # 89237-464
SmartCal™ Moisture Analyzer Test Substance
SmartCal™ Moisture Analyzer Test Substance
Moisture analyzer test system (set of 12)
Price
$11,569.23
Log in Para disponibilidad
Each (1g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
SmartCal™ is an innovative test substance that provides a fast way to verify the performance of a moisture analyzer.
  • Simply pour SmartCal™ into the sample pan and start
  • Test takes just 10 minutes
  • Easy to monitor and interpret instrument's performance over time
  • Store results directly in the instrument or manually enter into a free validated Excel® report sheet
  • Done just like a normal moisture measurement
  • Certified version available to satisfy regulatory requirements
  • SmartCal™ to get you up and running right away
SmartCal™ is a granular test substance. Its high sensitivity reliably detects adverse influences from ambient conditions and inaccurate heating or weighing devices.
Testing regularly with SmartCal™ ensures that moisture measurements are reliable at all times.
Documents
Especificaciones
  • Description:
    Moisture analyzer test system (set of 12)