Orders LinkContactUsLinkComponent
SmartCal™ Moisture Analyzer Test Substance
Moisture analyzer test system, certified version (set of 12)
  89237-460
 :  30005793
SmartCal™ Moisture Analyzer Test Substance
  89237-460
SmartCal™ Moisture Analyzer Test Substance
SmartCal™ Moisture Analyzer Test Substance
Moisture analyzer test system, certified version (set of 12)
$7,074.01
Each (1g)
SmartCal™ is an innovative test substance that provides a fast way to verify the performance of a moisture analyzer.
  • Simply pour SmartCal™ into the sample pan and start
  • Test takes just 10 minutes
  • Easy to monitor and interpret instrument's performance over time
  • Store results directly in the instrument or manually enter into a free validated Excel® report sheet
  • Done just like a normal moisture measurement
  • Certified version available to satisfy regulatory requirements
  • SmartCal™ to get you up and running right away
SmartCal™ is a granular test substance. Its high sensitivity reliably detects adverse influences from ambient conditions and inaccurate heating or weighing devices.
Testing regularly with SmartCal™ ensures that moisture measurements are reliable at all times.
  • Description:
    Moisture analyzer test system, certified version (set of 12)