Order Entry
Mexico
Orders LinkContactUsLinkComponent
Protector® Premier® Laboratory Hoods, Labconco®

Protector® Premier® Laboratory Hoods, Labconco®

Protector® Premier® Laboratory Hood without Blower, 115 V, 5', 0, 0
Catalog # 89232-748
Product available on GSA Advantage®
   Sustainable Materials|Energy Efficient
Supplier #:  100500000
Protector® Premier® Laboratory Hoods, Labconco®
Catalog # 89232-748
Supplier:  
Product available on GSA Advantage®
   Sustainable Materials|Energy Efficient
Protector® Premier® Laboratory Hoods, Labconco®
Protector® Premier® Laboratory Hoods, Labconco®

Protector® Premier® Laboratory Hood without Blower, 115 V, 5', 0, 0

Product Details & Documents

Labconco Protector® Premier® Fiberglass Fume Hoods have a one-piece liner of specially-formulated, fiberglass-reinforced polyester, which offers corrosion and fire resistance and easy clean up. There are no seams where deterioration can occur or where contaminants can collect. These general purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.

Hoods are available with a built-in standard fiberglass blower, or an explosion-proof fiberglass blower, or without a blower (remote blower sold separately).

Hoods are available with a built-in standard fiberglass blower, or an explosion-proof fiberglass blower, or without a blower (remote blower sold separately).

There are no seams where deterioration can occur or where contaminants can collect. These general purpose hoods are ideal for corrosive fumes but should not be used with perchloric acid, or radioactive or biological hazards.

 : All models are ETL listed. 230V models are CE listed.

 : Customize your hood with optional service fixtures that accommodate air, gas, vacuum or water lines. Complete your fume hood with a variety of accessories including blowers, work surfaces, storage cabinets, fire extinguishers, and air flow monitors (sold separately).

 : VWRCATALYST™ offers customer on-site validation, testing and certification services including face velocity profile test, smoke generation test, tracer gas test, IQ, OQ, and PQ. Please call 888-793-2300 or email VWR[email protected] for more details. You can also visit https://vwr.com/equip_services.

Especificaciones
  • Description:
    Protector® Premier® Laboratory Hood without Blower
  • Electrical:
    115 V
  • Nominal Width:
    5'
  • Service Fixtures:
    0
  • Electrical Duplex Outlet:
    0
  • Cat. No.:
    89232-748
  • Environmentally Preferable:
    Sustainable Materials|Energy Efficient
Accessories

Información del producto

  • Catalog #
    Availability
    Price
  • 4' SpillStopper™ Work Surface, Right Side Cupsink Cutout
    4' SpillStopper™ Work Surface, Right Side Cupsink Cutout
    89232-800
    Log in Para disponibilidad
    Each (1UN) - $41,340.00

Información del producto

  • Catalog #
    Availability
    Price
  • 4' SpillStopper™ Work Surface, No Cupsink Cutouts
    4' SpillStopper™ Work Surface, No Cupsink Cutouts
    89232-792
    Log in Para disponibilidad
    Each (1UN) - $36,860.00