Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
RIBOZOL RNA EXTR REAGENT
Catalog # MP97064-952
Supplier:
VWR
Certificates
Expand image
RIBOZOL RNA EXTR REAGENT
Catalog # MP97064-952
Supplier:
VWR
Supplier Number:
N580-30ML
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Additional Product Details
VWR GHS Compliance Letter (US)
Documentation
Frequently Bought Together
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International