Polysorbate 80 NF, Multi-Compendial, J.T.Baker®
Glass bottle, amber, narrow mouth, 1 L
Supplier: AVANTOR PERFORMANCE MATERIALS MX
: 4117-06
Polysorbate 80 NF, Multi-Compendial, J.T.Baker®
Catalog # JT4117-6
Supplier: AVANTOR PERFORMANCE MATERIALS MX
CAS Number: 9005-65-6
Jump to:
Product Details & Documents
Vegetable based.
- GMP manufactured product
- Vegetable based
Suitable for use in the manufacture of parenteral dosage forms.
Sinónimos: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
- CAS Number:9005-65-6
- Número MDL:MFCD00082107
- Punto de fusión:100 °C
- Densidad:1.062 g/cm³
- :Management of Change (MOC) category = R
- Punto de fusión:–21 °C
- Punto de inflamación:113 °C (closed cup)
- Temperatura de almacenamiento:Ambient
- EINECS:500-019-9
Especificaciones
Resultados de prueba de especificación
- NF - Acid Value:≤ 2.0
- NF - Hydroxyl Value:65 - 80
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Residue on Ignition:≤ 0.25 %
- NF - Saponification Value:45 - 55
- NF - Specific Gravity at 25°C:1.06 - 1.09
- NF - Viscosity at 25°C, cSt:300 - 500
- NF - Water (H₂O):≤ 3.0 %
- NF - Peroxide Value:≤ 10.0
- NF - Ethylene Oxide:≤ 1 ppm
- NF - Dioxane:≤ 10 ppm
- NF - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- NF - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- NF - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- NF - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- NF - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- NF - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- NF - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- EP - Acid Value:≤ 2.0
- EP - Total Ash:≤ 0.25 %
- EP - Hydroxyl Value:65 - 80
- EP - Identification A:Passes Test
- EP - Identification D:Passes Test
- EP - Peroxide Value:≤ 10.0
- EP - Ethylene Oxide:≤ 1 ppm
- EP - Dioxan:≤ 10 ppm
- EP - Saponification Value:45 - 55
- EP - Water (H₂O):≤ 3.0 %
- EP/BP - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- EP/BP - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- EP/BP - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- EP/BP - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- EP/BP - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- EP/BP - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- EP/BP - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- Appearance:Passes Test
- JP - Acid Value:≤ 2.0
- JP - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- JP - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- JP - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- JP - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- JP - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- JP - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- JP - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- JP - Dioxane:≤ 10 ppm
- JP - Ethylene Oxide:≤ 1 ppm
- JP - Heavy Metals (as Pb):≤ 20 ppm
- JP - Hydroxyl Value, meq KOH/g:65 - 80
- JP - Identification:Passes Test
- JP - Peroxide Value:≤ 10.0
- JP - Residue on Ignition:≤ 0.25 %
- JP - Water (H₂O):≤ 3.0 %
- JP - Saponification Value:45 - 55
- Additional Tests - Color (Gardner):≤ 7
- Additional Tests - Odor (Faint):Passes Test
- Additional Tests - Peroxide Value, meqO₂/kg:≤ 2.0
- Additional Tests - Endotoxin Concentration (EU/mL):≤ 10
- Free Ethylene Oxide:≤ 1 ppm
- 1,4-Dioxane:≤ 5 ppm
- Microbiological - Total Aerobic Microbial Count (cfu/g):≤ 100
- Microbiological - Escherichia Coli:Passes Test
- Microbiological - Pseudomonas aeruginosa:Passes Test
- Microbiological - Salmonella:Passes Test
- Microbiological - Staphylococcus aureus:Passes Test
- Microbiological - Total Yeast and Mold Count (cfu/g):≤ 50
- Ethylene glycol:≤ 0.10 %
- Diethylene glycol :≤ 0.10 %
Especificaciones
- Pack type:Glass bottle, amber, narrow mouth
- Size:1 L
Especificaciones
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



