Orders LinkContactUsLinkComponent
VWR® Maximum/Minimum Thermometers
VWR® Maximum/Minimum Thermometers
Organic Liquid, −35…+50 °C (−30…+120 °F), 200 mm, 1 °C/2 °F, 1 °C at ambient, 2 °C at –30 °C tolerance between limbs 1.5 °C
Catalog # 98000-178
VWR® Maximum/Minimum Thermometers
Catalog # 98000-178
Supplier:  
VWR® Maximum/Minimum Thermometers
VWR® Maximum/Minimum Thermometers
Organic Liquid, −35…+50 °C (−30…+120 °F), 200 mm, 1 °C/2 °F, 1 °C at ambient, 2 °C at –30 °C tolerance between limbs 1.5 °C
Discontinued
Product Details & Documents
Organic liquid thermometer features a U-shaped tube mounted on a plastic casing with metal floats at the top of each column.
  • Easy-to-understand dial thermometer
  • Minimum and maximum temperature floats are reset easily
One float remains at the maximum temperature while the other float remains at the minimum temperature
Dial thermometer displays the current temperature and registers the maximum and minimum temperatures attained since the last reset.
Reset the floats by pushing the reset button.
Especificaciones
  • Description:
    Organic Liquid
  • Dimensions:
    200 mm
  • Divisions:
    1 °C/2 °F
  • Accuracy:
    1 °C at ambient, 2 °C at –30 °C tolerance between limbs 1.5 °C
  • Range:
    −35…+50 °C (−30…+120 °F)