Order Entry
Mexico
Orders LinkContactUsLinkComponent
VWR® Water Sampling Kits

VWR® Water Sampling Kits

540 ml (18 oz.), 3 mil., 114×229 mm (4.5×9")
Catalog # 89003-818
Supplier:   VWR International
VWR® Water Sampling Kits
Catalog # 89003-818
Supplier:  VWR International
VWR® Water Sampling Kits
VWR® Water Sampling Kits

540 ml (18 oz.), 3 mil., 114×229 mm (4.5×9")

Price
$2,166.16
Log in for availability
Case (100UN)
Product Details & Documents

These sterile sampling bags are ideal for collecting chlorinated water specimens.

  • Sterile polyethylene construction
  • Suited for water sampling
  • Easy to use
  • Wire closure provides an airtight seal

They are made of sterile virgin polyethylene tubing with no side seals.

Bags feature a wire closure for an airtight seal and wide-sealed bottoms for added leakproof protection. They are easy to use: simply tear off perforated top, pull tabs to open bag, fill, grab ends of tape closure and pull shut, twirl bag three or four times to seal, and bend ends of closure inward to lock the seal.

UPA-approved for drinking water analysis.

Bags include a nontoxic, nutritive tablet containing 10 mg of active sodium thiosulfate.

Specifications
  • Capacity:
    540 ml (18 oz.)
  • Thickness:
    3 mil.
  • W×L:
    114×229 mm (4.5×9")