Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

VWR® Ring Supports, Laboratory

210 mm (81/4"), 12 L, 150 mm (6")
  56250-104
   Low Manufacturing Impact
VWR® Ring Supports, Laboratory
  56250-104
   Low Manufacturing Impact
VWR® Ring Supports, Laboratory
VWR® Ring Supports, Laboratory

210 mm (81/4"), 12 L, 150 mm (6")

 
$1,110.39
   
Paquete de 3
 

Ring supports are made of compressed cork for supporting flasks or dishes.

  • Compressed cork construction
  • Internal bevel at top
  • Solid support for round-bottomed vessels

An internal bevel at top provides a secure support for round-bottomed vessels.

 
  • Ext.Ø:
    210 mm (81/4")
  • Inner Diameter:
    150 mm (6")
  • For Flask Size:
    12 L
  • Cat. No.:
    56250-104
  • Environmentally Preferable:
    Low Manufacturing Impact