STERI-Buffer®, Veltek Associates
Phosphate buffer solution, 99 mL (3.3 oz.)
: SB100-99
Developed for use as a buffer solution and diluent in lab testing such a bioburden testing, pharmaceutical microbial limits testing, and AET testing and as a rinsing agent."
Sealed with a “no tamper” strip. Filtered at 0.2µm. Gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation.
- Description:Phosphate buffer solution
- Size:99 mL (3.3 oz.)