Orders LinkContactUsLinkComponent
STERI-Buffer®, Veltek Associates
Phosphate buffer solution, 99 mL (3.3 oz.)
  14222-662
 :  SB100-99
STERI-Buffer®, Veltek Associates
  14222-662
 :  
STERI-Buffer®, Veltek Associates
STERI-Buffer®, Veltek Associates
Phosphate buffer solution, 99 mL (3.3 oz.)
 :  
$12,752.03
Case (72UN)
Developed for use as a buffer solution and diluent in lab testing such a bioburden testing, pharmaceutical microbial limits testing, and AET testing and as a rinsing agent."
Sealed with a “no tamper” strip. Filtered at 0.2µm. Gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation.
  • Description:
    Phosphate buffer solution
  • Size:
    99 mL (3.3 oz.)