Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

HISTOSETTE® I Processing and Embedding Cassettes, Simport Scientific

Tissue Cassettes, Fluorescent Yellow
  87002-410
 :  M490-18
HISTOSETTE® I Processing and Embedding Cassettes, Simport Scientific
  87002-410
HISTOSETTE® I Processing and Embedding Cassettes, Simport Scientific
HISTOSETTE® I Processing and Embedding Cassettes, Simport Scientific

Tissue Cassettes, Fluorescent Yellow

 
$4,409.30
   
Case (1,500UN)
 
   
 

These disposable plastic cassettes with lids are made of acetal polymer and hold tissue or biopsy specimens during the embedding process and for storage. They keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The lid can be easily snapped apart and locked into the base of the cassette. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassette. The anterior writing area is at a 30° angle. The biopsy cassettes feature 1mm square openings.

They keep specimens safely submerged in liquid and are totally resistant to the chemical action of histological solvents. The efficient flow-through slots maximize fluid exchange and ensure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. The lid can be easily snapped apart and locked into the base of the cassette. Lids can be opened and closed as often as necessary and always relock securely without danger of specimen loss. Large labeling areas are located on three sides of the cassette. The anterior writing area is at a 30° angle. The biopsy cassettes feature 1mm square openings.

 : Cassettes are available in 11 non cytotoxic, non metallic colors, and three fluorescent colors. Each case contains three dispenser boxes of 500 cassettes.

 

 
  • Color:
    Fluorescent Yellow
  • Description:
    Tissue Cassettes

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.