Order Entry
Mexico
Orders LinkContactUsLinkComponent
Polysorbate 80, J.T.Baker®

Polysorbate 80, J.T.Baker®

Glass bottle, amber, narrow mouth, 500 ml
Catalog # JTX257-7
Supplier #:  X257-07
Polysorbate 80, J.T.Baker®
Catalog # JTX257-7
CAS Number:  9005-65-6
MOC:  Non-RegulatedCertificates
Polysorbate 80, J.T.Baker®
Polysorbate 80, J.T.Baker®

Glass bottle, amber, narrow mouth, 500 ml

CAS Number:  9005-65-6
MOC:  Non-Regulated
Price
$3,116.42
Log in for availability
Each (500ml) Case de 12 (500ml)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Polyoxyethylene (20) Sorbitan Monooleate

Synonyms:  Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
  • CAS Number:9005-65-6
  • MDL Number:MFCD00082107
  •  :Management of Change (MOC) category = N
  • Storage Temperature:Ambient
  • EINECS:500-018-3

Documents

Certificates
Search by Lot Number
Specifications

Specification Test Results

  • Identification (by IR):
    Passes Test

Specifications

  • Pack type:
    Glass bottle, amber, narrow mouth
  • Size:
    500 ml

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.