Orders LinkContactUsLinkComponent
Viscometer Holders, Cannon
Viscometer Holders, Cannon
Ubbelohde Removeable Viscometer Holder, Metal
Catalog # 47735-288
Supplier #:  9726-M82
Viscometer Holders, Cannon
Catalog # 47735-288
Viscometer Holders, Cannon
Viscometer Holders, Cannon
Ubbelohde Removeable Viscometer Holder, Metal
Price
$7,600.67
Log in for availability
Each (1UN)
Product Details & Documents
All CANNON viscometer holders fit holes 51 mm (2" ) in diameter.
  • Self-aligning
  • Constructed of chrome-plated machined brass
  • All metal holders are provided with a handle
Plastic holders are self-aligning and hold the viscometer firmly by means of a spring clip. Rubber holders are made of neoprene rubber.
Specifications
  • Description:
    Ubbelohde Removeable Viscometer Holder
  • Material:
    Metal