Order Entry
Mexico
Orders LinkContactUsLinkComponent
Orion™ pH Buffer Bottles, Thermo Scientific

Orion™ pH Buffer Bottles, Thermo Scientific

pH 9.18 buffer, 475 ml, Colorless
Catalog # 10010-640
Número de proveedor #:  910918
Orion™ pH Buffer Bottles, Thermo Scientific
Catalog # 10010-640
Supplier:  
CAS Number:  
Orion™ pH Buffer Bottles, Thermo Scientific
Orion™ pH Buffer Bottles, Thermo Scientific

pH 9.18 buffer, 475 ml, Colorless

CAS Number:  
Discontinued
El producto ha sido descontinuado. Sin embargo, encontramos productos alternativos similares.
Alternativas
BDH BUFFER PH 9.18 CLRLS 500ML Catalog #BDH5066-500ML

El producto BDH5066-500ML es un reemplazo para 10010-640 y tiene el mismo embalaje y especificaciones, pero puede tener una cantidad diferente.

Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

Perform accurate and reproducible pH calibrations with Orion™ pH buffer bottles.

  • All bottles feature pH/temperature tables printed on the labels for accurate calibrations and NIST traceability
  • Manufactured under ISO 9000 quality standards
  • Available in pint (475 ml) bottles or portable five packs of 2 oz. (5×60 ml) bottles

The pH 4.01, 5.00, 7.00, and 10.01 buffers are color-coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.

Documents

Especificaciones

Especificaciones

  • Color:
    Colorless
  • Description:
    pH 9.18 buffer
  • Size:
    475 ml

Especificaciones

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.