Powder. For determination of potassium. Lot analysis on label.
- Lot analysis on label
: Sodium cobaltnitrite, Trisodium hexanitritocobaltate
- :13600-98-1
- :Na₃Co(NO₂)₆
- :MFCD00003512
- :403.94 g/mol
- :13,08677
- :~ 220 °C
- :∼220 °C
- :Ambient
- :237-077-7
- :1479
- :5.1,III
- :Management of Change (MOC) category = N
- :5.1,II
Documents
3656 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
- Insoluble Matter:≤0.02 %
- Suitability for Potassium Determination:Passes Test
- Pack type:Poly bottle, white, blue T/E cap
- Size:500 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



