Agar for bacteriology
2.5 kg
Supplier: VWR
Número de proveedor #: J637-2.5KG
Jump to:
Product Details & Documents
A solidifying agent used in bacteriology. Agar consists of unbranched polysaccharides of galactose subunits derived from algae or seaweed.
- Agar for the growth of bacteria and fungi
- Typically used at concentrations of 1-2% in solid media preparation
Sinónimos: Agar Agar
- CAS Number:9002-18-0
- Número MDL:MFCD00081288
- Índice Merck:13,00184
- Punto de fusión:90 °C
- Temperatura de almacenamiento:Ambient
- EINECS:232-658-1
Documents
Certificates
Buscar por número de loteEspecificaciones
Especificaciones
- Size:2.5 kg



