Order Entry
Mexico
Orders LinkContactUsLinkComponent
Avantor® ACE® Buffers

Avantor® ACE® Buffers

1 L
Catalog # 97063-504
Supplier:   VWR
Número de proveedor #:  K249-1L
Avantor® ACE® Buffers
Catalog # 97063-504
Supplier:  VWR
CAS Number:  
Avantor® ACE® Buffers
Avantor® ACE® Buffers

1 L

CAS Number:  
Price
$2,221.37
Log in Para disponibilidad
Each (1l)
Product Details & Documents

These optimized buffers for DNA capillary electrophoresis sequencing provide excellent results, long read lengths, high resolution, and consistent run times

  • Long Read Lengths
  • High Resolution
  • Consistent Run Times

A.C.E.™ 10X Sequencing Buffer concentrate is conveniently diluted to the desired working concentration, thereby providing an easy and cost-effective buffer for your operating systems. VWR® A.C.E.™ 1X Sequencing Buffer comes ready-to-use, no dilution required, offering convenience and time savings. The tinted buffers have a slight pink hue in order to help the user differentiate between VWR® A.C.E.™ buffer and any other water, buffer, or solution in the sequencer. Tinted A.C.E.™ buffers have the same high-quality formulation as all A.C.E.™ products, delivering consistent and reproducible results.

Documents

Certificates
Buscar por número de lote
Especificaciones

Especificaciones

  • Size:
    1 L

Especificaciones