Orders LinkContactUsLinkComponent
Agarose I™, AMRESCO
Agarose I™, AMRESCO
500 g
Catalog # 97062-250
Supplier:   VWR
Número de proveedor #:  0710-500G
Agarose I™, AMRESCO
Catalog # 97062-250
Supplier:  VWR
Agarose I™, AMRESCO
Agarose I™, AMRESCO
500 g
Price
$5,642.11
Log in Para disponibilidad
Each (500g) Case de 12 (500g)
Product Details & Documents
Agarose I™ is for routine nucleic acid analytical/preparative applications.
  • All-Purpose, High-Purity Agarose
  • Exceptional Band Resolution and Clarity
  • Nuclease- and Protease-free
A standard melting/gelling agarose, suitable for routine nucleic acid analytical/preparative applications. It has a low EEO for shorter electrophoretic runs without compromising resolution. Excellent for blotting techniques and general manipulations.
Especificaciones
  • Size:
    500 g