Orders LinkContactUsLinkComponent
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
BD BBL™ Eosin Methylene Blue Agar, Levine, 500 g
Catalog # 90000-122
Número de proveedor #:  211221
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
Catalog # 90000-122
CAS Number:  
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
BD BBL™ Eosin Methylene Blue Agar, Levine, 500 g
CAS Number:  
Price
$5,290.00
Log in Para disponibilidad
Each (500g)
Product Details & Documents
This dehydrated culture media is used for isolating and differentiating lactose-fermenting from lactose-nonfermenting gram-negative enteric bacilli.
Documents
Especificaciones
Especificaciones
  • Description:
    BD BBL™ Eosin Methylene Blue Agar, Levine
  • Size:
    500 g
Especificaciones