BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
BD BBL™ Eosin Methylene Blue Agar, Levine, 500 g
Supplier: BECTON DICKINSON DE MEXICO, S. MX
Número de proveedor #: 211221
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
Catalog # 90000-122
Supplier: BECTON DICKINSON DE MEXICO, S. MX
CAS Number:
Jump to:
Product Details & Documents
This dehydrated culture media is used for isolating and differentiating lactose-fermenting from lactose-nonfermenting gram-negative enteric bacilli.
Documents
Certificates
Buscar por número de loteEspecificaciones
Especificaciones
- Description:BD BBL™ Eosin Methylene Blue Agar, Levine
- Size:500 g