Orders LinkContactUsLinkComponent
Straight Plug Rubber Stoppers, Wheaton
Straight Plug Rubber Stoppers, Wheaton
Mouth with ID×OD 13×20 mm, Straight plug rubber stoppers, durameter 45, Gray chlorobutyl
Catalog # 89168-268
Supplier #:  W224100-173
Straight Plug Rubber Stoppers, Wheaton
Catalog # 89168-268
Straight Plug Rubber Stoppers, Wheaton
Straight Plug Rubber Stoppers, Wheaton
Mouth with ID×OD 13×20 mm, Straight plug rubber stoppers, durameter 45, Gray chlorobutyl
Price
$11,781.00
Log in Para disponibilidad
Case (1,000UN)
Product Details & Documents
Wheaton straight plug stoppers provide maximum sealing for vials with a straight-wall glass finish
Stoppers are autoclavable.
Stoppers are available in a variety of sizes and rubber formulations . Straight plug stoppers are manufactured from the highest quality raw materials and are manufactured to yield exceptional dimensional stability.
Especificaciones
  • Description:
    Straight plug rubber stoppers, durameter 45
  • Material:
    Gray chlorobutyl
  • Fits:
    Mouth with ID×OD 13×20 mm