Straight Plug Rubber Stoppers, Wheaton
Mouth with ID×OD 13×20 mm, Straight plug rubber stoppers, durameter 45, Gray chlorobutyl
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Supplier #: W224100-173
Straight Plug Rubber Stoppers, Wheaton
Catalog # 89168-268
Supplier: DWK LIFE SCIENCES S. DE R.L. DE MX
Jump to:
Product Details & Documents
Wheaton straight plug stoppers provide maximum sealing for vials with a straight-wall glass finish
Stoppers are autoclavable.
Stoppers are available in a variety of sizes and rubber formulations . Straight plug stoppers are manufactured from the highest quality raw materials and are manufactured to yield exceptional dimensional stability.
Documents
Especificaciones
- Description:Straight plug rubber stoppers, durameter 45
- Material:Gray chlorobutyl
- Fits:Mouth with ID×OD 13×20 mm