Orders LinkContactUsLinkComponent
BD GasPak™ EZ Container Systems, BD Diagnostics
BD GasPak™ EZ Container Systems, BD Diagnostics
Incubation Container Rack, Standard
Catalog # 89024-786
Número de proveedor #:  260673
BD GasPak™ EZ Container Systems, BD Diagnostics
Catalog # 89024-786
BD GasPak™ EZ Container Systems, BD Diagnostics
BD GasPak™ EZ Container Systems, BD Diagnostics
Incubation Container Rack, Standard
Price
$4,930.00
Log in Para disponibilidad
Each (1UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Rapid gas generating sachets produce optimal bacterial growth in anaerobic, microaerophilic (reduced oxygen), or CO2 enriched environments.
Pouch systems include 20 resealable pouches and 20 rapid gas-generating sachets. The Anaerobe Pouch System also includes 20 dry anaerobic indicators. Sachets do not require water or a catalyst to produce optimal bacterial growth in anaerobic, microaerophilic (reduced oxygen), or CO2 enriched environments. Pouches are one-step resealable; no sealing bars needed. Multi-layer plastic pouch material preserves environmental integrity for maximum performance. Pouches can accommodate up to four Petri dishes.
Sachets do not require water or a catalyst. Two incubation container sizes are available: standard and large. These containers are nonbreakable and chemical-resistant for maximum safety. Easy-close latches assure a secure, airtight seal. Containers are designed to hold either petri dish racks or standard test tube racks for greater convenience. Standard container and rack holds 15 to 18 petri dishes and the large container and rack holds 30 to 33 petri dishes.
Ordering information: When using the anaerobic system, an anaerobic indicator must be used. Dry indicator strips change from blue to colorless in the absence of oxygen and may be stored at room temperature prior to use.
Especificaciones
  • Description:
    Incubation Container Rack, Standard
Accessories
Información del producto
  • Catalog #
    Availability
    Price
  • Anaerobic Indicator Strips, Dry, Pack of 100
    Anaerobic Indicator Strips, Dry, Pack of 100
    90003-866
    Log in Para disponibilidad
    Paquete de 100 - $4,470.00
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.