Orders LinkContactUsLinkComponent
Embedding Cassettes, Simport Scientific
Embedding Cassettes, Simport Scientific
Embedding Cassettes, Lilac
Catalog # 82028-094
Supplier #:  M480-10
Embedding Cassettes, Simport Scientific
Catalog # 82028-094
Embedding Cassettes, Simport Scientific
Embedding Cassettes, Simport Scientific
Embedding Cassettes, Lilac
Price
$3,483.75
Log in for availability
Case (1,500UN)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These disposable plastic tissue cassettes are suitable for holding and identifying tissue samples in processing, embedding, and sectioning procedures. The cassettes fit securely in microtome chuck adapters. They are molded from a high-density acetal polymer that is totally resistant to the chemical action of histological solvents, except for clear cassette 82028-100, which is made of polypropylene. The cassettes are designed to accept standard metal lids and will keep specimens safe during processing. The 45° anterior, slanted writing surface accepts markings easily, permitting sample identification throughout all stages of embedding. The cassettes can also be used for automatic cassette labeling instruments.
The cassettes fit securely in microtome chuck adapters. They are molded from a high-density acetal polymer that is totally resistant to the chemical action of histological solvents, except for clear cassette 82028-100, which is made of polypropylene. The cassettes are designed to accept standard metal lids and will keep specimens safe during processing. The 45° anterior, slanted writing surface accepts markings easily, permitting sample identification throughout all stages of embedding. The cassettes can also be used for automatic cassette labeling instruments.
Ordering information: Cassettes are packed in three dispenser boxes of 500 cassettes.
Specifications
  • Color:
    Lilac
  • Description:
    Embedding Cassettes
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Metal Lid
    Metal Lid
    87002-382
    Log in for availability
    Case (25UN) - $2,352.29