Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Alginic acid NF

Fiber drum, 25 kg
Catalog # 700013-072
 :  AL172-25KGBL
Alginic acid NF
Catalog # 700013-072
CAS Number:  9005-32-7
Alginic acid NF

Fiber drum, 25 kg

CAS Number:  9005-32-7
Price
$117,258.34
Log in Para disponibilidad
Each (25kg)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents

ALGINIC ACID,NF, is also referred to as algin or alginate, occurs naturally in the cell walls of brown algae. As it binds with water it creates a viscous gum-like texture. When dried it is capable of absorbing 200 to 300 times its own weight in water. It is often used in slimming aid products and in the manufacture of paper and textiles, as well as, being added to drinks, ice-creams and cosmetics as a thickening agent. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP). 

  • CAS Number:9005-32-7
  • Fórmula:(C₆H₈O₆)n
  • Número MDL:MFCD00081309
  • Molecular Weight:176.13 g/mol
  • Temperatura de almacenamiento:Ambient
Especificaciones

Resultados de prueba de especificación

  • Acid Value (Dried Basis):
    230 min.
  • pH:
    1.5 - 3.5
  • Microbial Limits:
    To Pass Test
  • Loss On Drying:
    max. 15.0 %
  • Total Ash:
    max. 4.0 %

Especificaciones

  • Pack type:
    Fiber drum
  • Size:
    25 kg

Especificaciones

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment.