Prefilter for Masks, 3M™
N95 filter for use with 3M™ 5000 series respirators, 6000 series cartridges, and prefilter retainer 56222-826, For use with 3M™ 5000 series respirators, 6000 series cartridges, and prefilter retainer 56222-826
Catalog # 56221-054
Supplier: 3M OCCUPATIONAL HEALTH
: 7000002008
Jump to:
Product Details & Documents
For use with 5000-series respirators.
- NIOSH 42 CFR 84 approved
: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Especificaciones
- Description:N95 filter for use with 3M™ 5000 series respirators, 6000 series cartridges, and prefilter retainer 56222-826
- Approved For Use Against:For use with 3M™ 5000 series respirators, 6000 series cartridges, and prefilter retainer 56222-826