Nunc® CryoColor Coder Inserts, Thermo Scientific
Purple
Supplier: Thermo Fisher Scientific
Supplier #: 375922
Nunc® CryoColor Coder Inserts, Thermo Scientific
Catalog # 24270-118
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
For comprehensive sample identification, fits inside CryoTube® vial cap.
- Made of polystyrene
- Available in 10 different colors
: Assortment 24270-120 includes either 50 of each color (for a pack) or 200 of each color (for a case).
Especificaciones
- Color:Purple