Orders LinkContactUsLinkComponent
VWR® RNase Removing Solution, Nucleaseliminator
NUCLEASELIMINATOR™, 500 mL
  97063-076
 :   VWR
 :  E891-500ML
VWR® RNase Removing Solution, Nucleaseliminator
  97063-076
 :  VWR
 :  
VWR® RNase Removing Solution, Nucleaseliminator
VWR® RNase Removing Solution, Nucleaseliminator
NUCLEASELIMINATOR™, 500 mL
 :  
$1,718.18
Each (500ml)
DNase and RNase contamination ruins expensive samples and time-consuming protocols in research laboratories every day. NucleasEliminator™ can help protect your investment by removing and deactivating even high concentrations of nuclease contamination from glass or plastic surfaces.
  • Effectively removes and deactivates nuclease contamination even at high concentrations
  • Safe on all laboratory glass and plastic ware
  • Stable at room temperature for at least two years
Simply wipe, immerse or fill the surface or vessel to be decontaminated with NucleasEliminator™, incubate 5 minutes at room temperature and rinse thoroughly with nuclease-free water. NucleasEliminator™ is safe to use on all surfaces at the laboratory bench ranging from glassware, microcentrifuge tubes, pipette tips and electrophoresis units. NucleasEliminator™ is supplied as a ready-to-use liquid. A 100 ml bottle will treat up to 100 microcentrifuge tubes or 25 agarose electrophoresis casting trays.
  • Description:
    NUCLEASELIMINATOR™
  • Size:
    500 mL