Order Entry
Mexico
Orders LinkContactUsLinkComponent
Electrode Fill Solutions

Electrode Fill Solutions

3M KCl saturated with AgCl, Bottle, 25 ml, —
Catalog # 97001-714
Supplier #:  51343184
Electrode Fill Solutions
Catalog # 97001-714
CAS Number:  
Electrode Fill Solutions
Electrode Fill Solutions

3M KCl saturated with AgCl, Bottle, 25 ml, —

CAS Number:  
Price
$990.29
Log in for availability
Each (25ml)
Product Details & Documents

FRISCOLYT-B™ (84.4% glycerol, 11% KCl, and 4.6% H₂O) is for low temperature samples.

Delivery information: Supplied in 25 ml (0.9 oz.) bottles.

Documents

Specifications

Specifications

  • Pack type:
    Bottle
  • Description:
    3M KCl saturated with AgCl
  • Ion:
  • Volume:
    25 ml

Specifications