Order Entry
Mexico
Orders LinkContactUsLinkComponent
Difco™ EC Medium, BD Diagnostics

Difco™ EC Medium, BD Diagnostics

EC Medium with MUG, 500 g
Catalog # 90001-606
Número de proveedor #:  222200
Difco™ EC Medium, BD Diagnostics
Catalog # 90001-606
CAS Number:  
Difco™ EC Medium, BD Diagnostics

EC Medium with MUG, 500 g

CAS Number:  
Price
$18,840.00
Log in Para disponibilidad
Each (500g)
Product Details & Documents

Differentiation and enumeration of fecal and nonfecal coliforms in water, wastewater, shellfish and foods.

Documents

Especificaciones

Especificaciones

  • Description:
    EC Medium with MUG
  • Size:
    500 g

Especificaciones

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.