Order Entry
Mexico
Orders LinkContactUsLinkComponent
Difco™ Fluid Lactose Medium, Dehydrated, BD Diagnostics

Difco™ Fluid Lactose Medium, Dehydrated, BD Diagnostics

Difco™ Fluid lactose medium, 500 g
Catalog # 90000-392
Supplier #:  211835
Difco™ Fluid Lactose Medium, Dehydrated, BD Diagnostics
Catalog # 90000-392
CAS Number:  
Difco™ Fluid Lactose Medium, Dehydrated, BD Diagnostics
Difco™ Fluid Lactose Medium, Dehydrated, BD Diagnostics

Difco™ Fluid lactose medium, 500 g

CAS Number:  
Price
$3,110.00
Log in for availability
Each (500g)
Product Details & Documents

Difco™ Fluid Lactose Medium is used in microbiology for the cultivation of Salmonella and coliform organisms in water, food, dairy and pharmaceutical products.

  • Salmonella cultivation
  • Coliform organism cultivation

Documents

Specifications

Specifications

  • Description:
    Difco™ Fluid lactose medium
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.