Order Entry
Mexico
Orders LinkContactUsLinkComponent
 

Benstat™ Divider Tote Boxes, Metro™

27.5 L (0.97 cu.ft.), 11 long, 15 short, 39.7W×52.4L×14.9H cm (155/8×205/8×57/8"), 44.5W×57.2L×15.2H cm (171/2×221/2×6")
Catalog # 82002-536
Supplier:   Metro
 :  TB93060BAS
Benstat™ Divider Tote Boxes, Metro™
Catalog # 82002-536
Supplier:  Metro
Benstat™ Divider Tote Boxes, Metro™
Benstat™ Divider Tote Boxes, Metro™

27.5 L (0.97 cu.ft.), 11 long, 15 short, 39.7W×52.4L×14.9H cm (155/8×205/8×57/8"), 44.5W×57.2L×15.2H cm (171/2×221/2×6")

Price
$2,540.00
Log in Para disponibilidad
Each (1UN) Paquete de 6 (1UN)
Product Details & Documents

Tote boxes offer safe and efficient storage of items. Boxes are durable, stackable, and feature high impact strength. There is a smooth area on each side of the box that is a convenient place to affix labels and bar codes.

Insert style covers fit inside the rim of the tote box and feature a finger hole that enables easy removal. Card holders are also available. Choose from holders made from Benstat™ or vinyl.

Insert style covers fit inside the rim of the tote box and feature a finger hole that enables easy removal. Card holders are also available. Choose from holders made from Benstat™ or vinyl.

Boxes are durable, stackable, and feature high impact strength. There is a smooth area on each side of the box that is a convenient place to affix labels and bar codes.

Especificaciones
  • Capacity:
    27.5 L (0.97 cu.ft.)
  • Interior Dimensions:
    39.7W×52.4L×14.9H cm (155/8×205/8×57/8")
  • Exterior Dimensions:
    44.5W×57.2L×15.2H cm (171/2×221/2×6")
  • Max. No. of Dividers:
    11 long, 15 short
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.