Prefilter for Masks, 3M™
P100 filter "organic vapor odors, solid and liquid particles including oil" for use with 3M™ 5000, 6000, 7000 series respirators, Removes organic vapor odors below the OSHA PELs, 99.97% efficient at removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, 7000 series respirators
Catalog # 56621-940
Supplier: 3M OCCUPATIONAL HEALTH
: 7000029657
Jump to:
Product Details & Documents
For use with 5000-series respirators.
- NIOSH 42 CFR 84 approved
: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Especificaciones
- Description:P100 filter "organic vapor odors, solid and liquid particles including oil" for use with 3M™ 5000, 6000, 7000 series respirators
- Approved For Use Against:Removes organic vapor odors below the OSHA PELs, 99.97% efficient at removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, 7000 series respirators