Prefilter for Masks, 3M™
P100 filter "solid and liquid particles" for use with 3M™ 5000, 6000, and 7000 series respirators., Provides 99.97% efficiency in removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, and 7000 series respirators
Catalog # 56222-931
Marketsource
Supplier: GRAINGER,S.A. DE C.V. MX
: 2091
Jump to:
Product Details & Documents
For use with 5000-series respirators.
- NIOSH 42 CFR 84 approved
: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
Especificaciones
- Description:P100 filter "solid and liquid particles" for use with 3M™ 5000, 6000, and 7000 series respirators.
- Approved For Use Against:Provides 99.97% efficiency in removing solid and liquid particles including those containing oil, use with 3M™ 5000, 6000, and 7000 series respirators