Orion™ pH Buffer Bottles, Thermo Scientific
pH 12.46 buffer, 475 ml, Colorless
Supplier: Thermo Fisher Scientific
: 910112
Orion™ pH Buffer Bottles, Thermo Scientific
Catalog # 10010-634
Supplier: Thermo Fisher Scientific
CAS Number:
Jump to:
Product Details & Documents
Perform accurate and reproducible pH calibrations with Orion™ pH buffer bottles.
- All bottles feature pH/temperature tables printed on the labels for accurate calibrations and NIST traceability
- Manufactured under ISO 9000 quality standards
- Available in pint (475 ml) bottles or portable five packs of 2 oz. (5×60 ml) bottles
The pH 4.01, 5.00, 7.00, and 10.01 buffers are color-coded for ease of use. All Orion™ pH buffers are traceable to NIST standard reference materials.
Certificates
Buscar por número de loteEspecificaciones
Especificaciones
- Color:Colorless
- Description:pH 12.46 buffer
- Size:475 ml



