Orders LinkContactUsLinkComponent
VWR® Tryptone
VWR® Tryptone
1 kg
Catalog # 97063-388
Supplier:   VWR
Número de proveedor #:  J859-1KG
VWR® Tryptone
Catalog # 97063-388
Supplier:  VWR
CAS Number:  
VWR® Tryptone
VWR® Tryptone
1 kg
CAS Number:  
Price
$4,072.95
Log in Para disponibilidad
Each (1kg) Case de 4 (1kg)
Product Details & Documents
High-purity pancreatic protein digests of casein used for media supplements.
  • High-purity
  • Pancreatic protein digests
  • Used for media supplements
High-purity pancreatic protein digests of casein used for media supplements.
Documents
Especificaciones
Especificaciones
  • Size:
    1 kg