Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
FOSFATO POTASIO MONOBASIC ANH
Catalog # 97063-348
Supplier:
VWR
Certificates
Expand image
FOSFATO POTASIO MONOBASIC ANH
Catalog # 97063-348
Supplier:
VWR
Supplier Number:
N854-600UL
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Datasheet
DFU-N854
Product Literature
VWR Life Science AMRESCO People-Safe Planet-Friendly Research Products Brochure
Documentation
Frequently Bought Together
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH