Order Entry
Mexico
Orders LinkContactUsLinkComponent
D-(+)-Glucose for biotechnology

D-(+)-Glucose for biotechnology

500 g
Catalog # 97061-168
Supplier:   VWR
Número de proveedor #:  0188-500G
D-(+)-Glucose for biotechnology
Catalog # 97061-168
Supplier:  VWR
CAS Number:  50-99-7
D-(+)-Glucose for biotechnology
D-(+)-Glucose for biotechnology

500 g

CAS Number:  50-99-7
Price
$1,061.01
Log in Para disponibilidad
Each (500g) Case de 12 (500g) Paleta de 288 (500g)
Product Details & Documents
Sinónimos:  Corn sugar, ᴅ-(+)-Glucose, Glucose, D-(+)-Dextrose, (2R,3S,4R,5R)-2,3,4,5,6-Pentahydroxyhexanal, Glucolin, Dextrose, D-Glucose
  • CAS Number:50-99-7
  • Fórmula:C₆H₁₂O₆
  • Número MDL:MFCD00148912
  • Molecular Weight:180.16 g/mol
  • Índice Merck:14,04459
  • Punto de fusión:146 °C
  • Densidad:1.54 g/cm³ (20 °C)
  • Temperatura de almacenamiento:Ambient
  • EINECS:200-075-1
Especificaciones

Resultados de prueba de especificación

  • Arsenic:
    = 0.00004%
  • Chloride:
    = 0.01%
  • DNase:
    none detected
  • Heavy Metals (as Pb):
    = 0.0005%
  • Insolubles:
    = 0.005%
  • Iron:
    = 0.0005%
  • Loss on Drying:
    = 0.2%
  • Protease:
    none detected
  • Purity:
    = 99.5%
  • Residue on Ignition:
    = 0.02%
  • RNase:
    none detected
  • Specific Rotation:
    52.5°- 54 °
  • Sulfate & Sulfite:
    = 0.005%

Especificaciones

  • Size:
    500 g

Especificaciones

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.