Order Entry
Mexico
Orders LinkContactUsLinkComponent
NIST Traceable pH Buffers with Temperature Table

NIST Traceable pH Buffers with Temperature Table

Bottle, 500 ml, 10.00
Catalog # 97007-788
Número de proveedor #:  63056129
NIST Traceable pH Buffers with Temperature Table
Catalog # 97007-788
CAS Number:  
NIST Traceable pH Buffers with Temperature Table
NIST Traceable pH Buffers with Temperature Table

Bottle, 500 ml, 10.00

CAS Number:  
Price
$1,761.57
Log in Para disponibilidad
Each (500ml)
Product Details & Documents

NIST-traceable buffer for pH electrode calibration and verification.

  • Temperature table on bottle

Measurement results depend on accurate calibrations, and accurate calibrations come from high quality solutions. Each standard solution comes with a certificate of analysis, providing traceability to NIST standards.

Documents

Especificaciones

Especificaciones

  • Pack type:
    Bottle
  • Size:
    500 ml
  • pH:
    10.00

Especificaciones

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.