Inoculum Broth, BD
B12 Inoculum Broth, 100 g
: 254210
Inoculum Broth is used for preparing the inoculum of lactobacilli and other microorganisms used in microbiological assays of vitamins and amino acids.
- Used for cultivating lactobacilli
- For preparing the inoculum for microbiological assays
- Peptone provides nitrogen and amino acids
- Beige, homogenous appearance when dehydrated
Inoculum Broth is the recommended medium for preparation of lactobacilli inoculum for vitamin assays.
- Description:B12 Inoculum Broth
- Size:100 g