Order Entry
Mexico
Orders LinkContactUsLinkComponent
Difco™ EC Medium, BD Diagnostics

Difco™ EC Medium, BD Diagnostics

EC Medium, 500 g
Catalog # 90002-320
Supplier #:  231430
Difco™ EC Medium, BD Diagnostics
Catalog # 90002-320
CAS Number:  
Difco™ EC Medium, BD Diagnostics

EC Medium, 500 g

CAS Number:  
Price
$5,370.00
Log in for availability
Each (500g)
Product Details & Documents

Differentiation and enumeration of fecal and nonfecal coliforms in water, wastewater, shellfish and foods.

Documents

Specifications

Specifications

  • Description:
    EC Medium
  • Size:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.