Orders LinkContactUsLinkComponent
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
BD BBL™ Eosin Methylene Blue Agar, Modified (Holt-Harris & Teague), 1 lb.
Catalog # 90000-118
Número de proveedor #:  211215
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
Catalog # 90000-118
CAS Number:  
BD BBL™ Eosin Methylene Blue Agar, BD Diagnostics
BD BBL™ Eosin Methylene Blue Agar, Modified (Holt-Harris & Teague), 1 lb.
CAS Number:  
Price
$5,950.00
Log in Para disponibilidad
Each (1lb)
Product Details & Documents
This dehydrated culture media is used for isolating and differentiating lactose-fermenting from lactose-nonfermenting gram-negative enteric bacilli.
Documents
Especificaciones
Especificaciones
  • Description:
    BD BBL™ Eosin Methylene Blue Agar, Modified (Holt-Harris & Teague)
  • Size:
    1 lb.