$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
BALANZA PRECISION 0.1G 8200
82028-348
:
VALERIO VALDERRAMA NOE SV
BALANZA PRECISION 0.1G 8200
82028-348
:
VALERIO VALDERRAMA NOE SV
:
ED8201-CW
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International